Assay Details
| Gene Symbol: Fdps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501583.2 | XP_003501631.1 | 6 - 7 | 814 | 63 | |
| GenBank mRNA | JP040072.1 | - | 4 - 5 | 511 | 63 | |
More Information
Set Membership: |
|

Fdps
Genomic Map