Assay Details
| Gene Symbol: Slc25a38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003505944.2 | XP_003505992.2 | 5 - 6 | 948 | 58 | |
| XM_007646432.1 | XP_007644622.1 | 6 - 7 | 807 | 58 | ||
More Information
Set Membership: |
|

Slc25a38
Genomic Map