Assay Details
| Gene Symbol: Nqo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003513191.2 | XP_003513239.1 | 4 - 5 | 472 | 67 | |
| XM_007653502.1 | XP_007651692.1 | 4 - 5 | 697 | 67 | ||
| XM_007653503.1 | XP_007651693.1 | 4 - 5 | 547 | 67 | ||
| GenBank mRNA | JI892097.1 | - | 3 - 4 | 379 | 67 | |
| JP037572.1 | - | 3 - 4 | 379 | 67 | ||
More Information
Set Membership: |
|

Nqo2
Genomic Map