Assay Details
| Gene Symbol: Ccbe1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007647553.1 | XP_007645743.1 | 4 - 5 | 344 | 56 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccbe1| Gene Symbol: Ccbe1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007647553.1 | XP_007645743.1 | 4 - 5 | 344 | 56 | |
Set Membership: |
|