Assay Details
| Gene Symbol: Fam92a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007648393.1 | XP_007646583.1 | 1 - 2 | 358 | 100 | |
| GenBank mRNA | JI865732.1 | - | 1 - 2 | 358 | 100 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam92a1| Gene Symbol: Fam92a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007648393.1 | XP_007646583.1 | 1 - 2 | 358 | 100 | |
| GenBank mRNA | JI865732.1 | - | 1 - 2 | 358 | 100 | |