Assay Details
| Gene Symbol: Il15ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003494915.2 | XP_003494963.1 | 4 - 5 | 698 | 62 | |
| GenBank mRNA | JI865468.1 | - | 4 - 5 | 698 | 62 | |
| JP063086.1 | - | 4 - 5 | 698 | 62 | ||
More Information
Set Membership: |
|

Il15ra
Genomic Map