Assay Details
| Gene Symbol: Slco3a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495276.1 | XP_003495324.1 | 1 - 2 | 492 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slco3a1| Gene Symbol: Slco3a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495276.1 | XP_003495324.1 | 1 - 2 | 492 | 65 | |
Set Membership: |
|