Assay Details
| Gene Symbol: Wsb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495907.2 | XP_003495955.1 | 1 - 2 | 252 | 59 | |
| GenBank mRNA | JI871456.1 | - | 1 - 2 | 212 | 59 | |
More Information
Set Membership: |
|

Wsb2
Genomic Map