Assay Details
| Gene Symbol: Rab8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499337.2 | XP_003499385.1 | 2 - 3 | 257 | 103 | |
| GenBank mRNA | JI895045.1 | - | 2 - 3 | 216 | 103 | |
| JP038970.1 | - | 2 - 3 | 216 | 103 | ||
More Information
Set Membership: |
|

Rab8b
Genomic Map