Assay Details
| Gene Symbol: LOC100755074 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500128.2 | XP_003500176.1 | 1 - 2 | 172 | 70 | |
| XM_007640849.1 | XP_007639039.1 | 2 - 3 | 363 | 70 | ||
| XM_007640850.1 | XP_007639040.1 | 1 - 2 | 168 | 70 | ||
| XM_007640851.1 | XP_007639041.1 | 2 - 3 | 227 | 70 | ||
| XM_007640852.1 | XP_007639042.1 | 2 - 3 | 213 | 70 | ||
More Information
Set Membership: |
|

LOC100755074
Genomic Map