Assay Details
| Gene Symbol: LOC100755074 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500128.2 | XP_003500176.1 | 3 - 4 | 797 | 78 | |
| XM_007640849.1 | XP_007639039.1 | 4 - 5 | 988 | 78 | ||
| XM_007640850.1 | XP_007639040.1 | 3 - 4 | 793 | 78 | ||
| XM_007640851.1 | XP_007639041.1 | 4 - 5 | 852 | 78 | ||
| XM_007640852.1 | XP_007639042.1 | 4 - 5 | 838 | 78 | ||
More Information
Set Membership: |
|

LOC100755074
Genomic Map