Assay Details
| Gene Symbol: Snrpd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503007.2 | XP_003503055.1 | 3 - 4 | 445 | 89 | |
| GenBank mRNA | JI885169.1 | - | 3 - 4 | 413 | 89 | |
| JP059897.1 | - | 3 - 4 | 413 | 89 | ||
More Information
Set Membership: |
|

Snrpd1
Genomic Map