Assay Details
| Gene Symbol: LOC100752747 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003505275.2 | XP_003505323.2 | 2 - 3 | 437 | 110 | |
| XM_007645796.1 | XP_007643986.1 | 3 - 4 | 331 | 110 | ||
| XM_007645797.1 | XP_007643987.1 | 2 - 3 | 437 | 110 | ||
More Information
Set Membership: |
|

LOC100752747
Genomic Map