Assay Details
| Gene Symbol: LOC100752747 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003505275.2 | XP_003505323.2 | 4 - 5 | 597 | 89 | |
| XM_007645796.1 | XP_007643986.1 | 5 - 6 | 491 | 89 | ||
| XM_007645797.1 | XP_007643987.1 | 4 - 5 | 597 | 89 | ||
More Information
Set Membership: |
|

LOC100752747
Genomic Map