Assay Details
| Gene Symbol: Mtfr1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510658.2 | XP_003510706.1 | 3 - 4 | 893 | 71 | |
| XM_007651268.1 | XP_007649458.1 | 3 - 4 | 897 | 71 | ||
| GenBank mRNA | JI863050.1 | - | 3 - 4 | 424 | 71 | |
| JP040213.1 | - | 3 - 4 | 394 | 71 | ||
More Information
Set Membership: |
|

Mtfr1l
Genomic Map