Assay Details
| Gene Symbol: LOC100751227 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003515145.2 | XP_003515193.2 | 2 - 3 | 286 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100751227| Gene Symbol: LOC100751227 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003515145.2 | XP_003515193.2 | 2 - 3 | 286 | 76 | |
Set Membership: |
|