Assay Details
| Gene Symbol: LOC100757335 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007647448.1 | XP_007645638.1 | 7 - 8 | 862 | 102 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100757335| Gene Symbol: LOC100757335 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007647448.1 | XP_007645638.1 | 7 - 8 | 862 | 102 | |
Set Membership: |
|