Assay Details
| Gene Symbol: LOC100762026 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500945.2 | XP_003500993.2 | 11 - 12 | 3012 | 66 | |
| GenBank mRNA | JI891705.1 | - | 10 - 11 | 2796 | 66 | |
| JP041251.1 | - | 10 - 11 | 2795 | 66 | ||
More Information
Set Membership: |
|

LOC100762026
Genomic Map