Assay Details
| Gene Symbol: LOC100762026 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500945.2 | XP_003500993.2 | 12 - 13 | 3138 | 66 | |
| GenBank mRNA | JI891705.1 | - | 11 - 12 | 2922 | 66 | |
| JP041251.1 | - | 11 - 12 | 2921 | 66 | ||
More Information
Set Membership: |
|

LOC100762026
Genomic Map