Assay Details
| Gene Symbol: LOC100762026 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500945.2 | XP_003500993.2 | 2 - 3 | 420 | 66 | |
| XM_007641662.1 | XP_007639852.1 | 2 - 3 | 420 | 66 | ||
More Information
Set Membership: |
|

LOC100762026
Genomic Map