Assay Details
| Gene Symbol: LOC100762026 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500945.2 | XP_003500993.2 | 4 - 5 | 647 | 100 | |
| XM_007641662.1 | XP_007639852.1 | 4 - 5 | 647 | 100 | ||
| GenBank mRNA | JI891705.1 | - | 3 - 4 | 431 | 100 | |
| JP041251.1 | - | 3 - 4 | 430 | 100 | ||
More Information
Set Membership: |
|

LOC100762026
Genomic Map