Assay Details
| Gene Symbol: Pdk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003505691.2 | XP_003505739.1 | 10 - 11 | 1298 | 64 | |
| XM_007646161.1 | XP_007644351.1 | 10 - 11 | 1298 | 64 | ||
| GenBank mRNA | JI865083.1 | - | 10 - 11 | 1081 | 64 | |
| JP065833.1 | - | 10 - 11 | 1081 | 64 | ||
More Information
Set Membership: |
|

Pdk3
Genomic Map