Assay Details
| Gene Symbol: Idh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506911.2 | XP_003506959.1 | 2 - 3 | 562 | 63 | |
| GenBank mRNA | JI872108.1 | - | 2 - 3 | 562 | 63 | |
| JP047295.1 | - | 2 - 3 | 561 | 63 | ||
More Information
Set Membership: |
|

Idh1
Genomic Map