Assay Details
| Gene Symbol: Mogs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003511143.2 | XP_003511191.2 | 2 - 3 | 266 | 64 | |
| GenBank mRNA | AB115503.1 | - | 1 - 2 | 223 | 64 | |
| JI876134.1 | - | 1 - 2 | 205 | 64 | ||
| JP051877.1 | - | 1 - 2 | 205 | 64 | ||
More Information
Set Membership: |
|

Mogs
Genomic Map