Assay Details
| Gene Symbol: Cnga4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003513168.2 | XP_003513216.1 | 6 - 7 | 1433 | 77 | |
| XM_007653493.1 | XP_007651683.1 | 3 - 4 | 1041 | 77 | ||
More Information
Set Membership: |
|

Cnga4
Genomic Map