Assay Details
| Gene Symbol: LOC103159292 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645952.1 | XP_007644142.1 | 2 - 3 | 689 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC103159292| Gene Symbol: LOC103159292 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645952.1 | XP_007644142.1 | 2 - 3 | 689 | 79 | |
Set Membership: |
|