Assay Details
| Gene Symbol: Rfx8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007640565.1 | XP_007638755.1 | 8 - 9 | 816 | 65 | |
| XM_007640566.1 | XP_007638756.1 | 11 - 12 | 1053 | 65 | ||
More Information
Set Membership: |
|

Rfx8
Genomic Map