Assay Details
| Gene Symbol: LOC100755657 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007641059.1 | XP_007639249.1 | 14 - 15 | 2902 | 87 | |
| XM_007641060.1 | XP_007639250.1 | 15 - 16 | 3361 | 87 | ||
| GenBank mRNA | JP051547.1 | - | 1 - 2 | 56 | 87 | |
More Information
Set Membership: |
|

LOC100755657
Genomic Map