Assay Details
| Gene Symbol: LOC100755657 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007641059.1 | XP_007639249.1 | 16 - 17 | 3133 | 93 | |
| XM_007641060.1 | XP_007639250.1 | 17 - 18 | 3592 | 93 | ||
| GenBank mRNA | JP051547.1 | - | 3 - 4 | 287 | 93 | |
More Information
Set Membership: |
|

LOC100755657
Genomic Map