Assay Details
| Gene Symbol: LOC100755657 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007641059.1 | XP_007639249.1 | 19 - 20 | 4778 | 108 | |
| XM_007641060.1 | XP_007639250.1 | 20 - 21 | 5237 | 108 | ||
| GenBank mRNA | JP051547.1 | - | 6 - 7 | 1932 | 108 | |
More Information
Set Membership: |
|

LOC100755657
Genomic Map