Assay Details
| Gene Symbol: LOC100755657 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007641059.1 | XP_007639249.1 | 20 - 21 | 4853 | 66 | |
| XM_007641060.1 | XP_007639250.1 | 21 - 22 | 5312 | 66 | ||
| GenBank mRNA | JP051547.1 | - | 7 - 8 | 2007 | 66 | |
More Information
Set Membership: |
|

LOC100755657
Genomic Map