Assay Details
| Gene Symbol: Ikbip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509492.2 | XP_003509540.1 | 1 - 2 | 249 | 85 | |
| XM_003509493.2 | XP_003509541.1 | 1 - 2 | 249 | 85 | ||
More Information
Set Membership: |
|

Ikbip
Genomic Map