Assay Details
| Gene Symbol: Ythdf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007626785.1 | XP_007624975.1 | 2 - 3 | 1738 | 69 | |
| GenBank mRNA | JI870626.1 | - | 2 - 3 | 1738 | 69 | |
More Information
Set Membership: |
|

Ythdf2
Genomic Map