Assay Details
| Gene Symbol: Nr2f6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007638694.1 | XP_007636884.1 | 2 - 3 | 701 | 90 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nr2f6| Gene Symbol: Nr2f6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007638694.1 | XP_007636884.1 | 2 - 3 | 701 | 90 | |
Set Membership: |
|