Assay Details
Gene Symbol: Wdr53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003495396.2 | XP_003495444.1 | 2 - 3 | 515 | 72 | |
XM_007640609.1 | XP_007638799.1 | 2 - 3 | 733 | 72 | ||
GenBank mRNA | JI866948.1 | - | 1 - 2 | 527 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
