Assay Details
| Gene Symbol: Slc25a10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496890.2 | XP_003496938.1 | 9 - 10 | 836 | 62 | |
| GenBank mRNA | JI863367.1 | - | 9 - 10 | 780 | 62 | |
| JP042069.1 | - | 9 - 10 | 780 | 62 | ||
More Information
Set Membership: |
|

Slc25a10
Genomic Map