Assay Details
| Gene Symbol: Arhgdib | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499207.2 | XP_003499255.1 | 5 - 6 | 521 | 68 | |
| XM_007639716.1 | XP_007637906.1 | 5 - 6 | 647 | 68 | ||
More Information
Set Membership: |
|

Arhgdib
Genomic Map