Assay Details
Gene Symbol: Mb21d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003503196.2 | XP_003503244.1 | 3 - 4 | 1192 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mb21d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003503196.2 | XP_003503244.1 | 3 - 4 | 1192 | 90 |
Set Membership: |
![]() ![]() ![]() ![]() |