Assay Details
| Gene Symbol: Pkdrej | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003513415.1 | XP_003513463.1 | 1 | 126 | 98 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pkdrej| Gene Symbol: Pkdrej | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003513415.1 | XP_003513463.1 | 1 | 126 | 98 | |
Set Membership: |
|