Assay Details
| Gene Symbol: Rcc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007626876.1 | XP_007625066.1 | 2 - 3 | 937 | 60 | |
| XM_007626882.1 | XP_007625072.1 | 2 - 3 | 936 | 60 | ||
More Information
Set Membership: |
|

Rcc1
Genomic Map