Assay Details
| Gene Symbol: Yes1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007653906.1 | XP_007652096.1 | 10 - 11 | 1432 | 77 | |
| GenBank mRNA | JI871006.1 | - | 2 - 3 | 295 | 77 | |
| JP039351.1 | - | 2 - 3 | 295 | 77 | ||
More Information
Set Membership: |
|

Yes1
Genomic Map