Assay Details
| Gene Symbol: Pbdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506985.2 | XP_003507033.2 | 2 - 3 | 228 | 70 | |
| GenBank mRNA | JI870794.1 | - | 2 - 3 | 192 | 70 | |
| JP044958.1 | - | 2 - 3 | 192 | 70 | ||
More Information
Set Membership: |
|

Pbdc1
Genomic Map