Assay Details
| Gene Symbol: LOC100755143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003508405.2 | XP_003508453.2 | 2 - 3 | 289 | 108 | |
| XM_007649034.1 | XP_007647224.1 | 3 - 4 | 299 | 108 | ||
| XM_007649035.1 | XP_007647225.1 | 3 - 4 | 420 | 108 | ||
More Information
Set Membership: |
|

LOC100755143
Genomic Map