Assay Details
| Gene Symbol: Mrps17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509486.2 | XP_003509534.1 | 2 - 3 | 219 | 78 | |
| GenBank mRNA | JI868465.1 | - | 2 - 3 | 226 | 78 | |
| JP042755.1 | - | 2 - 3 | 226 | 78 | ||
More Information
Set Membership: |
|

Mrps17
Genomic Map