Assay Details
| Gene Symbol: LOC100755143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JI876103.1 | - | 1 - 2 | 123 | 67 | |
| JP060431.1 | - | 3 - 4 | 458 | 67 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100755143| Gene Symbol: LOC100755143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JI876103.1 | - | 1 - 2 | 123 | 67 | |
| JP060431.1 | - | 3 - 4 | 458 | 67 | ||
Set Membership: |
|