Assay Details
| Gene Symbol: LOC100755143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JI876103.1 | - | 6 - 7 | 643 | 96 | |
| JP060431.1 | - | 8 - 9 | 978 | 96 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100755143| Gene Symbol: LOC100755143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JI876103.1 | - | 6 - 7 | 643 | 96 | |
| JP060431.1 | - | 8 - 9 | 978 | 96 | ||