Assay Details
| Gene Symbol: Tmem179b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007654452.1 | XP_007652642.1 | 2 - 3 | 388 | 57 | |
| XM_007654453.1 | XP_007652643.1 | 2 - 3 | 388 | 57 | ||
| GenBank mRNA | JI890537.1 | - | 1 - 2 | 191 | 57 | |
| JP065194.1 | - | 1 - 2 | 189 | 57 | ||
More Information
Set Membership: |
|

Tmem179b
Genomic Map