Assay Details
| Gene Symbol: Lats2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500425.2 | XP_003500473.1 | 3 - 4 | 688 | 73 | |
| XM_007641164.1 | XP_007639354.1 | 3 - 4 | 891 | 73 | ||
More Information
Set Membership: |
|

Lats2
Genomic Map