Assay Details
| Gene Symbol: Irg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503490.1 | XP_003503538.1 | 3 - 4 | 295 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Irg1| Gene Symbol: Irg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503490.1 | XP_003503538.1 | 3 - 4 | 295 | 66 | |
Set Membership: |
|