Assay Details
| Gene Symbol: Garem | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007647152.1 | XP_007645342.1 | 4 - 5 | 1493 | 98 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Garem| Gene Symbol: Garem | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007647152.1 | XP_007645342.1 | 4 - 5 | 1493 | 98 | |